siRNA Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 3 | 1161 | ||
| GenBank mRNA | AK014242.1 | - | 3 | 1039 | ||
| BC049157.1 | - | 3 | 1260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fezf1| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028462.1 | NP_082738.1 | 3 | 1161 | ||
| GenBank mRNA | AK014242.1 | - | 3 | 1039 | ||
| BC049157.1 | - | 3 | 1260 | |||