siRNA Details
| Gene Symbol: Xpot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081056.1 | NP_001074525.1 | 8 | 1068 | ||
| GenBank mRNA | AK029113.1 | - | 3 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpot| Gene Symbol: Xpot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081056.1 | NP_001074525.1 | 8 | 1068 | ||
| GenBank mRNA | AK029113.1 | - | 3 | 521 | ||