siRNA Details
| Gene Symbol: 3110043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081343.1 | NP_001074812.1 | 7 | 979 | ||
| GenBank mRNA | AK014175.1 | - | 6 | 431 | ||
| AK154817.1 | - | 7 | 953 | |||
| AK167354.1 | - | 7 | 910 | |||
| BC026738.1 | - | 5 | 384 | 4020832 | ||
| BC076612.1 | - | 7 | 1081 | 30638238 | ||

3110043O21Rik
Genomic Map