siRNA Details
| Gene Symbol: 3110082I17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028469.3 | NP_082745.2 | 5 | 1132 | ||
| GenBank mRNA | AK014271.1 | - | 5 | 1112 | ||
| BC044867.1 | - | 5 | 1104 | |||
| BC058580.1 | - | 4 | 1701 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110082I17Rik| Gene Symbol: 3110082I17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028469.3 | NP_082745.2 | 5 | 1132 | ||
| GenBank mRNA | AK014271.1 | - | 5 | 1112 | ||
| BC044867.1 | - | 5 | 1104 | |||
| BC058580.1 | - | 4 | 1701 | |||