siRNA Details
| Gene Symbol: Fam118a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133750.4 | NP_598511.1 | 3 | 260 | ||
| GenBank mRNA | AK082151.1 | - | 3 | 198 | ||
| AK143690.1 | - | 3 | 233 | |||
| AK180870.1 | - | 3 | 120 | |||
| BC016425.1 | - | 3 | 118 | 4487028 | ||
| BC058794.1 | - | 3 | 321 | 6486049 | ||
| BC066174.1 | - | 3 | 176 | 30109713 | ||
| CJ084461.1 | - | 3 | 261 | |||

Fam118a
Genomic Map