siRNA Details
| Gene Symbol: Fam118a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133750.4 | NP_598511.1 | 5 | 806 | ||
| GenBank mRNA | AK078807.1 | - | 3 | 544 | ||
| AK082151.1 | - | 5 | 744 | |||
| AK143690.1 | - | 5 | 779 | |||
| BC016425.1 | - | 5 | 664 | 4487028 | ||
| BC058794.1 | - | 5 | 867 | 6486049 | ||
| BC066174.1 | - | 5 | 722 | 30109713 | ||

Fam118a
Genomic Map