siRNA Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028472.2 | NP_082748.1 | 8 | 1316 | ||
| GenBank mRNA | AF454954.1 | - | 8 | 1316 | ||
| AK014221.1 | - | 6 | 521 | |||
| AY263358.1 | - | 8 | 837 | |||
| BC066153.1 | - | 8 | 821 | 30098453 | ||

Bmper
Genomic Map