siRNA Details
| Gene Symbol: Prl8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028477.2 | NP_082753.1 | 3, 4 | 317 | ||
| GenBank mRNA | AF525158.1 | - | 3, 4 | 274 | ||
| AK005485.1 | - | 3, 4 | 318 | |||
| AK014414.1 | - | 3, 4 | 318 | |||
| BC109185.1 | - | 3, 4 | 316 | 40050251 | ||
| BC109186.1 | - | 3, 4 | 316 | 40050253 | ||

Prl8a1
Genomic Map