siRNA Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028478.2 | NP_082754.2 | 2, 3 | 193 | ||
| GenBank mRNA | AK005472.1 | - | 2, 3 | 180 | ||
| AK132288.1 | - | 2, 3 | 194 | |||
| BC031808.1 | - | 2, 3 | 148 | |||
| BC039934.1 | - | 2, 3 | 204 | |||
| BC075725.1 | - | 2, 3 | 181 | |||

Rassf6
Genomic Map