siRNA Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028479.1 | NP_082755.1 | 6 | 896 | ||
| GenBank mRNA | AK038431.1 | - | 4 | 523 | ||
| AK217601.1 | - | 4 | 313 | |||
| BC042508.1 | - | 3 | 1702 | |||
| BC054431.1 | - | 4 | 792 | |||
| BC085320.1 | - | 2 | 2195 | |||
| BC110993.1 | - | 5 | 502 | 5363496 | ||
| BC116732.1 | - | 6 | 896 | 40126051 | ||
| BC116734.1 | - | 6 | 896 | 40126053 | ||

Mrgbp
Genomic Map