siRNA Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028479.1 | NP_082755.1 | 3 | 604 | ||
| GenBank mRNA | AK005551.1 | - | 2 | 244 | ||
| AK052575.1 | - | 2 | 259 | |||
| AK217601.1 | - | 1 | 21 | |||
| BC042508.1 | - | 2 | 218 | |||
| BC054431.1 | - | 2 | 191 | |||
| BC085320.1 | - | 1 | 710 | |||
| BC110993.1 | - | 2 | 213 | 5363496 | ||
| BC116732.1 | - | 3 | 604 | 40126051 | ||
| BC116734.1 | - | 3 | 604 | 40126053 | ||

Mrgbp
Genomic Map