siRNA Details
| Gene Symbol: Cib4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028483.1 | NP_082759.1 | 5, 6 | 500 | ||
| GenBank mRNA | AK006670.1 | - | 5, 6 | 501 | ||
| BC099558.1 | - | 5, 6 | 446 | |||
| BC118938.1 | - | 5, 6 | 465 | 40091772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cib4| Gene Symbol: Cib4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028483.1 | NP_082759.1 | 5, 6 | 500 | ||
| GenBank mRNA | AK006670.1 | - | 5, 6 | 501 | ||
| BC099558.1 | - | 5, 6 | 446 | |||
| BC118938.1 | - | 5, 6 | 465 | 40091772 | ||