siRNA Details
| Gene Symbol: Ddit4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030143.4 | NP_084419.2 | 3 | 526 | ||
| GenBank mRNA | AF327512.1 | - | 2 | 281 | ||
| AF335325.1 | - | 2 | 326 | |||
| AK006617.1 | - | 3 | 525 | |||
| AK018944.1 | - | 3 | 528 | |||
| AK029362.1 | - | 1 | 94 | |||
| AK053333.1 | - | 3 | 464 | |||
| AK075693.1 | - | 3 | 415 | |||
| BC038131.1 | - | 3 | 425 | |||
| BC147181.1 | - | 2 | 305 | 9056209 | ||
| BC147182.1 | - | 2 | 305 | 9056220 | ||

Ddit4l
Genomic Map