siRNA Details
| Gene Symbol: Ttc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183096.3 | NP_898919.3 | 12 | 1572 | ||
| GenBank mRNA | BC048386.1 | - | 12 | 1572 | 6310247 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc29| Gene Symbol: Ttc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183096.3 | NP_898919.3 | 12 | 1572 | ||
| GenBank mRNA | BC048386.1 | - | 12 | 1572 | 6310247 | |