siRNA Details
| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028500.3 | NP_082776.2 | 5 | 692 | ||
| NM_029782.3 | NP_084058.2 | 3 | 393 | |||
| GenBank mRNA | AB206786.1 | - | 5 | 597 | ||
| AK006582.1 | - | 5 | 693 | |||
| AK018263.1 | - | 3 | 315 | |||
| AK046081.1 | - | 3 | 394 | |||

Calr3
Genomic Map