siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163427.1 | NP_001156899.1 | 2 | 212 | ||
| NM_028505.2 | NP_082781.1 | 2 | 212 | |||
| GenBank mRNA | AK006658.1 | - | 2 | 178 | ||
| BC115452.2 | - | 2 | 158 | 40100110 | ||
| BG088874.2 | - | 2 | 211 | |||
| CJ075544.1 | - | 2 | 236 | |||

Pradc1
Genomic Map