siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163427.1 | NP_001156899.1 | 5 | 786 | ||
| NM_028505.2 | NP_082781.1 | 4 | 618 | |||
| GenBank mRNA | AK006658.1 | - | 5 | 752 | ||
| BC115452.2 | - | 4 | 564 | 40100110 | ||
| CJ208123.1 | - | 1 | 107 | |||

Pradc1
Genomic Map