siRNA Details
| Gene Symbol: Arhgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152801.2 | NP_690014.2 | 19 | 2206 | ||
| GenBank mRNA | AF393831.1 | - | 19 | 2206 | ||
| AK006635.1 | - | 12 | 1205 | |||
| BC019723.1 | - | 2 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef6| Gene Symbol: Arhgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152801.2 | NP_690014.2 | 19 | 2206 | ||
| GenBank mRNA | AF393831.1 | - | 19 | 2206 | ||
| AK006635.1 | - | 12 | 1205 | |||
| BC019723.1 | - | 2 | 333 | |||