siRNA Details
| Gene Symbol: Dcbld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028523.3 | NP_082799.2 | 16 | 2167 | ||
| GenBank mRNA | AF387548.1 | - | 16 | 2259 | ||
| AK006805.1 | - | 6 | 870 | |||
| AK051889.1 | - | 14 | 1445 | |||
| AK084652.1 | - | 5 | 998 | |||
| AK165117.1 | - | 4 | 1100 | |||
| BC066097.1 | - | 16 | 2161 | 6856458 | ||

Dcbld2
Genomic Map