siRNA Details
| Gene Symbol: Prss52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028525.2 | NP_082801.2 | 3 | 511 | ||
| GenBank mRNA | AB009660.1 | - | 3 | 548 | ||
| AK006746.1 | - | 3 | 510 | |||
| BC049714.1 | - | 3 | 511 | 6741678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss52| Gene Symbol: Prss52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028525.2 | NP_082801.2 | 3 | 511 | ||
| GenBank mRNA | AB009660.1 | - | 3 | 548 | ||
| AK006746.1 | - | 3 | 510 | |||
| BC049714.1 | - | 3 | 511 | 6741678 | ||