siRNA Details
| Gene Symbol: Nme8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167909.1 | NP_001161381.1 | 6 | 510 | ||
| NM_181591.3 | NP_853622.2 | 6 | 510 | |||
| GenBank mRNA | AF548543.1 | - | 6 | 502 | ||
| AK006819.1 | - | 6 | 502 | |||
| BC052356.1 | - | 6 | 510 | 6770702 | ||

Nme8
Genomic Map