siRNA Details
| Gene Symbol: Armt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024261.2 | NP_077223.2 | 2 | 148 | ||
| GenBank mRNA | AK006776.1 | - | 2 | 158 | ||
| AK038060.1 | - | 2 | 127 | |||
| AK134431.1 | - | 2 | 115 | |||
| BC002139.1 | - | 2 | 133 | |||
| BC099894.1 | - | 2 | 241 | |||
| BC138908.1 | - | 2 | 108 | 8861930 | ||
| BC145732.1 | - | 2 | 108 | 40131035 | ||
| BU148062.1 | - | 2 | 133 | |||

Armt1
Genomic Map