siRNA Details
| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028536.1 | NP_082812.1 | 3 | 888 | ||
| GenBank mRNA | AK006786.1 | - | 2 | 280 | ||
| AK086180.1 | - | 2 | 593 | |||
| BC024705.1 | - | 2 | 545 | 3668303 | ||
| BC036300.1 | - | 3 | 888 | 4501054 | ||

Ccsap
Genomic Map