siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 10 | 1194 | ||
| GenBank mRNA | AB093239.1 | - | 10 | 1138 | ||
| AK038491.1 | - | 10 | 1166 | |||
| AK087732.1 | - | 11 | 1352 | |||
| AK141925.1 | - | 10 | 1195 | |||
| AY196789.1 | - | 10 | 1193 | |||
| BC030362.1 | - | 10 | 1164 | 5369051 | ||

Hspa12a
Genomic Map