siRNA Details
| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018013.1 | NP_001018013.1 | 9 | 1193 | ||
| GenBank mRNA | AB195681.1 | - | 9 | 1193 | ||
| AK006830.1 | - | 9 | 1193 | |||
| BC139270.1 | - | 8 | 1396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Izumo1| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018013.1 | NP_001018013.1 | 9 | 1193 | ||
| GenBank mRNA | AB195681.1 | - | 9 | 1193 | ||
| AK006830.1 | - | 9 | 1193 | |||
| BC139270.1 | - | 8 | 1396 | |||