siRNA Details
| Gene Symbol: Kif2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028547.2 | NP_082823.1 | 1 | 2067 | ||
| GenBank mRNA | AK006866.1 | - | 1 | 367 | ||
| AK030188.1 | - | 1 | 2068 | |||
| BC100484.1 | - | 1 | 2043 | 30919380 | ||
| BC132056.1 | - | 1 | 2121 | 40130333 | ||
| BC137981.1 | - | 1 | 2121 | 8861000 | ||

Kif2b
Genomic Map