siRNA Details
| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028550.3 | NP_082826.1 | 4, 5 | 560 | ||
| GenBank mRNA | AK006951.1 | - | 3, 4 | 434 | ||
| AK030264.1 | - | 4, 5 | 560 | |||
| BC061035.1 | - | 3, 4 | 471 | 6774253 | ||

1700074P13Rik
Genomic Map