siRNA Details
| Gene Symbol: Mbd3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028557.2 | NP_082833.1 | 3 | 202 | ||
| GenBank mRNA | AK006927.1 | - | 3 | 203 | ||
| AK018931.1 | - | 3 | 197 | |||
| AY038023.1 | - | 2 | 151 | |||
| BC049738.1 | - | 3 | 218 | 6771508 | ||
| BC061034.1 | - | 3 | 208 | 6707322 | ||

Mbd3l1
Genomic Map