siRNA Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028526.4 | NP_082802.1 | 3 | 423 | ||
| NM_028560.4 | NP_082836.2 | 3 | 423 | |||
| GenBank mRNA | AK006750.1 | - | 3 | 424 | ||
| AK006964.1 | - | 3 | 414 | |||
| BC048621.1 | - | 3 | 401 | 6742909 | ||

Pebp4
Genomic Map