siRNA Details
| Gene Symbol: Speer4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028561.2 | NP_082837.1 | 5 | 1427 | ||
| GenBank mRNA | AK006975.1 | - | 5 | 1476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer4b| Gene Symbol: Speer4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028561.2 | NP_082837.1 | 5 | 1427 | ||
| GenBank mRNA | AK006975.1 | - | 5 | 1476 | ||