siRNA Details
| Gene Symbol: Tssk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183099.2 | NP_898922.2 | 2 | 556 | ||
| GenBank mRNA | AK007032.1 | - | 2 | 558 | ||
| BC114543.1 | - | 2 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tssk5| Gene Symbol: Tssk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183099.2 | NP_898922.2 | 2 | 556 | ||
| GenBank mRNA | AK007032.1 | - | 2 | 558 | ||
| BC114543.1 | - | 2 | 360 | |||