siRNA Details
Gene Symbol: Zfp433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243067.1 | NP_001229996.1 | 4 | 1234 | ||
GenBank mRNA | BC064812.1 | - | 4 | 1233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243067.1 | NP_001229996.1 | 4 | 1234 | ||
GenBank mRNA | BC064812.1 | - | 4 | 1233 |