siRNA Details
| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA427326.1 | - | 5 | 456 | ||
| AK008857.1 | - | 4 | 308 | |||
| AW105955.1 | - | 4 | 339 | |||
| BC021953.1 | - | 6 | 664 | |||
| BC099512.1 | - | 5 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210408F21Rik| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA427326.1 | - | 5 | 456 | ||
| AK008857.1 | - | 4 | 308 | |||
| AW105955.1 | - | 4 | 339 | |||
| BC021953.1 | - | 6 | 664 | |||
| BC099512.1 | - | 5 | 423 | |||