siRNA Details
| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA427326.1 | - | 6 | 576 | ||
| AK008857.1 | - | 6 | 482 | |||
| BC021953.1 | - | 8 | 838 | |||
| BC099512.1 | - | 7 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210408F21Rik| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA427326.1 | - | 6 | 576 | ||
| AK008857.1 | - | 6 | 482 | |||
| BC021953.1 | - | 8 | 838 | |||
| BC099512.1 | - | 7 | 597 | |||