siRNA Details
| Gene Symbol: Wdr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028599.2 | NP_082875.1 | 14 | 1495 | ||
| GenBank mRNA | AK010772.1 | - | 14 | 1495 | ||
| AK152412.1 | - | 14 | 1496 | |||
| AK163221.1 | - | 14 | 1469 | |||
| AK168163.1 | - | 14 | 1497 | |||
| AK169932.1 | - | 14 | 1493 | |||
| BC049925.1 | - | 3 | 227 | |||
| BC055483.1 | - | 3 | 396 | |||
| BC085177.1 | - | 14 | 1525 | |||
| BC117880.2 | - | 14 | 1525 | 40096010 | ||
| BC117881.1 | - | 14 | 1522 | 40096014 | ||

Wdr75
Genomic Map