siRNA Details
| Gene Symbol: Glipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028608.3 | NP_082884.1 | 3 | 711 | ||
| GenBank mRNA | AK010768.1 | - | 3 | 643 | ||
| AK137557.1 | - | 3 | 658 | |||
| AK170051.1 | - | 3 | 658 | |||
| BC025083.1 | - | 3 | 635 | 3711757 | ||
| BC098231.1 | - | 3 | 685 | 3325823 | ||

Glipr1
Genomic Map