siRNA Details
| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028611.3 | NP_082887.2 | 9 | 1086 | ||
| GenBank mRNA | AK010752.1 | - | 9 | 1085 | ||
| AK090097.1 | - | 9 | 1085 | |||
| AK184109.1 | - | 2 | 194 | |||
| BC028524.1 | - | 2 | 325 | |||
| BC108350.1 | - | 9 | 1073 | 4220568 | ||
| BC116213.1 | - | 2 | 178 | |||
| BC116214.1 | - | 2 | 178 | |||
| BC128473.1 | - | 2 | 178 | |||
| BC128474.1 | - | 2 | 178 | |||

Ndufaf7
Genomic Map