siRNA Details
| Gene Symbol: Dppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028615.1 | NP_082891.1 | 5 | 645 | ||
| GenBank mRNA | AF490346.1 | - | 5 | 443 | ||
| AK010743.1 | - | 5 | 443 | |||
| AK145645.1 | - | 5 | 472 | |||
| AK145760.1 | - | 5 | 442 | |||
| AK163375.1 | - | 5 | 445 | |||
| AK167211.1 | - | 5 | 443 | |||
| AY561246.1 | - | 5 | 645 | |||
| BC137768.1 | - | 5 | 448 | 8860787 | ||

Dppa2
Genomic Map