siRNA Details
| Gene Symbol: Dppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028615.1 | NP_082891.1 | 4 | 589 | ||
| GenBank mRNA | AF490346.1 | - | 4 | 387 | ||
| AK010743.1 | - | 4 | 387 | |||
| AK145645.1 | - | 4 | 416 | |||
| AK145760.1 | - | 4 | 386 | |||
| AK163375.1 | - | 4 | 389 | |||
| AK167211.1 | - | 4 | 387 | |||
| AY561246.1 | - | 4 | 589 | |||
| BC137768.1 | - | 4 | 392 | 8860787 | ||

Dppa2
Genomic Map