siRNA Details
| Gene Symbol: Rbm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170847.1 | NP_001164318.1 | 2 | 1299 | ||
| GenBank mRNA | AK142175.1 | - | 2 | 1300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm20| Gene Symbol: Rbm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170847.1 | NP_001164318.1 | 2 | 1299 | ||
| GenBank mRNA | AK142175.1 | - | 2 | 1300 | ||