siRNA Details
| Gene Symbol: Lce1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028622.2 | NP_082898.1 | 2 | 662 | ||
| GenBank mRNA | AK003705.1 | - | 2 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce1c| Gene Symbol: Lce1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028622.2 | NP_082898.1 | 2 | 662 | ||
| GenBank mRNA | AK003705.1 | - | 2 | 663 | ||