siRNA Details
| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 4, 5 | 332 | ||
| GenBank mRNA | AK003561.1 | - | 3, 4 | 291 | ||
| BC033045.1 | - | 3, 4 | 310 | 1068150 | ||
| BC094619.1 | - | 4, 5 | 319 | |||
| CJ085020.1 | - | 4, 5 | 347 | |||

Fcf1
Genomic Map