siRNA Details
| Gene Symbol: 1110008P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198001.2 | NP_932118.1 | 2 | 258 | ||
| GenBank mRNA | AK003598.1 | - | 2 | 213 | ||
| AK081091.1 | - | 216 | ||||
| BC024615.1 | - | 2 | 225 | 2812475 | ||
| BC049639.1 | - | 2 | 177 | 6590746 | ||
| BY264831.1 | - | 2 | 283 | |||

1110008P14Rik
Genomic Map