siRNA Details
| Gene Symbol: 1110034G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028637.1 | NP_082913.1 | 2, 3 | 633 | ||
| GenBank mRNA | AK004090.1 | - | 2, 3 | 207 | ||
| BC100475.1 | - | 2, 3 | 633 | 30253755 | ||
| BC138171.1 | - | 2, 3 | 287 | 8861190 | ||

1110034G24Rik
Genomic Map