siRNA Details
| Gene Symbol: 4930421P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015172.1 | - | 1 | 60 | ||
| AK077193.1 | - | 1 | 55 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930421P05Rik| Gene Symbol: 4930421P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015172.1 | - | 1 | 60 | ||
| AK077193.1 | - | 1 | 55 | |||