siRNA Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028662.2 | NP_082938.1 | 3 | 1131 | ||
| GenBank mRNA | AF414190.1 | - | 4 | 804 | ||
| AK090329.1 | - | 3 | 1320 | |||
| AK144403.1 | - | 4 | 827 | |||
| AK152362.1 | - | 4 | 805 | |||
| AK154868.1 | - | 4 | 790 | |||
| AK155551.1 | - | 4 | 837 | |||
| AK167343.1 | - | 3 | 1132 | |||
| BC025875.1 | - | 4 | 803 | 5137689 | ||
| BC036992.1 | - | 4 | 811 | 5003855 | ||

Slc35b2
Genomic Map