siRNA Details
| Gene Symbol: Ankrd45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028664.1 | NP_082940.1 | 4 | 702 | ||
| GenBank mRNA | AK016757.1 | - | 3 | 621 | ||
| BC049713.1 | - | 4 | 674 | 6741553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd45| Gene Symbol: Ankrd45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028664.1 | NP_082940.1 | 4 | 702 | ||
| GenBank mRNA | AK016757.1 | - | 3 | 621 | ||
| BC049713.1 | - | 4 | 674 | 6741553 | ||