siRNA Details
| Gene Symbol: 4930415O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201322.1 | NP_001188251.1 | 3 | 277 | ||
| GenBank mRNA | AK015155.1 | - | 3 | 261 | ||
| AK029626.1 | - | 3 | 261 | |||
| BC048564.1 | - | 3 | 277 | 6705104 | ||
 Back To Top
 Back To Top
 4930415O20Rik
4930415O20Rik Genomic Map
Genomic Map Back To Top
 Back To Top