siRNA Details
| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028672.2 | NP_082948.2 | 3 | 1337 | ||
| GenBank mRNA | AK040751.1 | - | 1 | 378 | ||
| AK133447.1 | - | 3 | 1353 | |||
| BC024460.1 | - | 4 | 1270 | 4975427 | ||
| BC026495.1 | - | 3 | 1337 | 4505506 | ||

Fam161a
Genomic Map