siRNA Details
| Gene Symbol: Zdbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267872.1 | NP_001254801.1 | 5 | 617 | ||
| NM_001285936.1 | NP_001272865.1 | 6 | 751 | |||
| NM_001285937.1 | NP_001272866.1 | 5 | 694 | |||
| NM_028673.1 | NP_082949.1 | 6 | 667 | |||
| GenBank mRNA | AB777269.1 | - | 5 | 694 | ||
| AB777270.1 | - | 6 | 751 | |||
| AK015271.1 | - | 6 | 668 | |||
| AK132808.1 | - | 5 | 618 | |||
| BC050985.1 | - | 228 | ||||

Zdbf2
Genomic Map